Simvastatin 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (1S,3R,7S,8S,8aR)-8-(2-((2R,4R)-4-Hydroxy-6-oxotetrahydro-2H-pyran-2-yl)ethyl)-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl 2,2-dimethylbutanoate

458840010 458840050 458840250
ACRO458840010EA 47.1 EUR
ACRO458840010 ACRO458840050 ACRO458840250
Simvastatin 98%
Simvastatin
Formel: C25H38O5
Molecular Weight: 418.57 g/mol
Schmelzpunkt: 127…132 °C
Flammpunkt: 127 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00072007
CAS-Nummer: 79902-63-9

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 125 to 135 °C
Loss on drying ≤0.5 %
HPLC ≥97.5 %

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR