2-Fluoroadenine 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 6-Amino-2-fluoropurine, 6-Amino-2-fluoro-7H-purine, 2-Fluoro-7H-purin-6-amine

459160010 459160050
ACRO459160010EA 154 EUR
ACRO459160010 ACRO459160050
2-Fluoroadenine 97%
2-Fluoroadenine
Formel: C₅H₄FN₅
Molecular Weight: 153.12 g/mol
Schmelzpunkt: >350 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01632749
CAS-Nummer: 700-49-2

Bestellen

Spezifikation Testergebnisse

Appearance White to brown Powder or crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR