2-Chlor-5-nitrophenol 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
459040050
459040250
ACRO459040050EA
54.4
EUR
ACRO459040050
ACRO459040250
2-Chlor-5-nitrophenol 98%
2-Chlor-5-nitrophenol
Formel:
C₆H₄ClNO₃ Molecular Weight: 173.56 g/mol Schmelzpunkt: 119…121 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01571825 CAS-Nummer: 619-10-3 |
Spezifikation Testergebnisse
Appearance | Off-white to brown Powder or solid |
Infrared spectrum | Conforms |
Melting point | 117 to 123 °C |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...