3,5-Dibromanilin 98%

Lieferant: Thermo Scientific
Achtung

460560050 460560250
ACRO460560050EA 68.5 EUR
ACRO460560050 ACRO460560250
3,5-Dibromanilin 98%
3,5-Dibromanilin
Formel: C₆H₅Br₂N
Molecular Weight: 250,92 g/mol
Schmelzpunkt: 53…54,5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00047841
CAS-Nummer: 626-40-4

Bestellen

Spezifikation Testergebnisse

Appearance Brown to black Solid or crystalline solid
Infrared spectrum Conforms
Melting point 51 to 56 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR