BAM15 ≥98%, kristalliner Feststoff
Lieferant: Cayman Chemical
Synonyme:
BAM 15, N5,N6-Bis(2-fluorophenyl)-[1,2,5]oxadiazolo[3,4-b]pyrazine-5,6-diamine
BAM15 is a mitochondrial protonophore uncoupler which increase proton transport into the mitochondrial matrix through an ATP synthase-independent pathway, thereby uncoupling nutrient oxidation from ATP production.
- High oxygen flux than FCCP
These are used to determine rate of cellular respiration and have antioxidant effects that protect from ischemia-reperfusion injury. The off target activity of this uncouplers leads to the undesired effects, including plasma membrane depolarization, mitochondrial inhibition, and cytotoxicity.
Formel:
C₁₆H₁₀F₂N₆O Molecular Weight: 340,29 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00373912 CAS-Nummer: 210302-17-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...