Mebendazol
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Methyl [5-(Benzoyl)benzimidazol-2-yl]carbamate, Methyl (5-benzoyl-1H-benzo[d]imidazol-2-yl)carbamate
101720050
101720250
101720000
ACRO101720050EA
30.9
EUR
ACRO101720050
ACRO101720250
101720000.
Mebendazol
Mebendazol
Formel:
C₁₆H₁₃N₃O₃ Molecular Weight: 295.30 g/mol Schmelzpunkt: 287 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00057872 CAS-Nummer: 31431-39-7 |
Spezifikation Testergebnisse
Appearance | White to off-white to grey Powder |
Infrared spectrum | Conforms |
Melting point | 285 to 290 °C |
Loss on drying | ≤0.5 % (1 g, 105 °C, 4 h) |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...