5-Hydroxyoxindole 96%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Hydroxyindolin-2-one

153550010 153550050
ACRO153550010EA 74.3 EUR
ACRO153550010 ACRO153550050
5-Hydroxyoxindole 96%
5-Hydroxyoxindole
Formel: C₈H₇NO₂
Molecular Weight: 149,15 g/mol
Schmelzpunkt: 266…272 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00014566
CAS-Nummer: 3416-18-0

Bestellen

Spezifikation Testergebnisse

Appearance White to brown Powder or crystalline powder
Infrared spectrum Conforms
HPLC ≥95.0 %
E/E 95 to 97 % v/v (calculated)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR