S-Tritylcystein 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: H-Cys(Trt)-OH

173010050 173010250
ACRO173010050EA 35.9 EUR
ACRO173010050 ACRO173010250
S-Tritylcystein 98%
S-Trityl-L-cysteine
Formel: C₂₂H₂₁NO₂S
Molecular Weight: 363,5 g/mol
Schmelzpunkt: 182…183 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002611
CAS-Nummer: 2799-07-7

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Granular powder
Infrared spectrum Conforms
Loss on drying ≤2 %
HPLC ≥97.5 %
Specific optical rotation +107° to +119° (20°C, 589 nm) (0.8% in 0.04M HCl (EtOH))

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR