Norethindrone

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Norethisterone

262890010 262890050
ACRO262890010EA 89.4 EUR
ACRO262890010 ACRO262890050
Norethindrone
Norethindrone
Formel: C₂₀H₂₆O₂
Molecular Weight: 298.426 g/mol
Schmelzpunkt: 205…206 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00067596
CAS-Nummer: 68-22-4

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 205 to 208 °C
Loss on drying ≤0.5 % (1 g, 105 °C, 3 h)
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR