Tetrakis(trimethylsilyl)silan 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: TMSS

293900010 293900050
ACRO293900010EA 34.3 EUR
ACRO293900010 ACRO293900050
Tetrakis(trimethylsilyl)silan 97%
Tetrakis(trimethylsilyl)silan
Formel: C₁₂H₃₆Si₅
Molecular Weight: 320,842 g/mol
Schmelzpunkt: >300 °C (Subl.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00054859
CAS-Nummer: 4098-98-0

Bestellen

Spezifikation Testergebnisse

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR