p-Toluenesulphonyl cyanide 95%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
4-Methylbenzenesulphonyl cyanide, Tosyl cyanide
455820010
455820050
ACRO455820010EA
49.6
EUR
ACRO455820010
ACRO455820050
p-Toluenesulphonyl cyanide 95%
p-Toluenesulphonyl cyanide
Formel:
C₈H₇NO₂S Molecular Weight: 181,21 g/mol Schmelzpunkt: 47…50 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00009977 CAS-Nummer: 19158-51-1 UN: 3261 ADR: 8,II |
Spezifikation Testergebnisse
Appearance | White to tan Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 47 to 50 °C |
HPLC | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...