4-Brom-o-toluylsäure 97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 4-Bromo-2-methylbenzoic acid

443400050 443400250
ACRO443400050EA 65 EUR
ACRO443400050 ACRO443400250
4-Brom-o-toluylsäure 97%
4-Brom-o-toluylsäure
Formel: C₈H₇BrO₂
Molecular Weight: 215,05 g/mol
Schmelzpunkt: 182…184 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00040905
CAS-Nummer: 68837-59-2
UN: 2811
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance White to light brown Powder or crystalline powder
Infrared spectrum Conforms
Melting point 180 to 185 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR