4-Thiazolcarbonsäure 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Thiazole-4-carboxylic acid

443970010 443970050
ACRO443970010EA 62.5 EUR
ACRO443970010 ACRO443970050
4-Thiazolcarbonsäure 97%
4-Thiazolecarboxylic acid
Formel: C₄H₃NO₂S
Molecular Weight: 129,1371 g/mol
Schmelzpunkt: 195…199 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00623589
CAS-Nummer: 3973-08-8

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 190 to 205 °C
Loss on drying ≤3 %
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR