4-Brom-2-methoxypyridin 97%
Lieferant: Thermo Scientific
|
Achtung
|
450000010
450000050
ACRO450000010EA
88.8
EUR
ACRO450000010
ACRO450000050
4-Brom-2-methoxypyridin 97%
4-Bromo-2-methoxypyridine
Formel:
C₆H₆BrNO Siedepunkt: 87 °C (15 mmHg) Lagertemperatur: Kühlschrank |
MDL:
MFCD08277268 CAS-Nummer: 100367-39-3 |
Spezifikation Testergebnisse
Appearance | Colorless to yellow Low melting solid or liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5540 to 1.5560 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...