5-Cyclopropyl-1,2,4-oxadiazol-3-amine 97%
Lieferant: Thermo Scientific
|
Achtung
|
450030010
450030050
ACRO450030010EA
201
EUR
ACRO450030010
ACRO450030050
5-Cyclopropyl-1,2,4-oxadiazol-3-amine 97%
5-Cyclopropyl-1,2,4-oxadiazol-3-amine
Formel:
C₅H₇N₃O Lagertemperatur: Raumtemperatur |
MDL:
MFCD11847813 CAS-Nummer: 868696-42-8 |
Spezifikation Testergebnisse
Appearance | White to yellow Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...