(Isocyanoimino)triphenylphosphorane 90%

Lieferant: Thermo Scientific
Achtung

452920050 452920010
ACRO452920050EA 218 EUR
ACRO452920050 ACRO452920010
(Isocyanoimino)triphenylphosphorane 90%
(Isocyanoimino)triphenylphosphorane
Formel: C₁₉H₁₅N₂P
Molecular Weight: 302,32 g/mol
Schmelzpunkt: 160…161 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09038528
CAS-Nummer: 73789-56-7

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow-orange Crystalline powder
Infrared spectrum Conforms
HPLC ≥88 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR