Clotrimazol
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-(2-Chlorotrityl)imidazole, 1-[(2-Chlorophenyl)diphenylmethyl]imidazole
455460050
455460250
455461000
ACRO455460050EA
69.4
EUR
ACRO455460050
ACRO455460250
ACRO455461000
Clotrimazol
Clotrimazole
Formel:
C₂₂H₁₇ClN₂ Molecular Weight: 344,84 g/mol Schmelzpunkt: 141…145 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00057220 CAS-Nummer: 23593-75-1 |
Spezifikation Testergebnisse
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Melting point | 142 to 152 °C |
Loss on drying | ≤0.5 % |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...