Arbutin 98%

Lieferant: Thermo Fisher Scientific

Synonyme: 4-Hydroxyphenyl beta-D-Glucopyranoside, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(4-hydroxyphenoxy)tetrahydro-2H-pyran-3,4,5-triol, Hydroquinone-beta-D-glucoside

455640050 455640250 455641000
ACRO455640050EA 46.1 EUR
ACRO455640050 ACRO455640250 ACRO455641000
Arbutin 98%
Arbutin
Formel: C₁₂H₁₆O₇
Molecular Weight: 272.25 g/mol
Schmelzpunkt: 195…201 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00016915
CAS-Nummer: 497-76-7

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 196 to 201 °C
Loss on drying ≤2 %
HPLC ≥97.5 %
Specific optical rotation -63° to -67° (20 °C, 589 nm) (c=3, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR