N-Formyl-L-proline 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: N-For-Pro-OH, (S)-1-Formylpyrrolidine-2-carboxylic acid, (S)-1-Formyl-2-pyrrolidinecarboxylic acid

456420010 456420050
ACRO456420010EA 70.5 EUR
ACRO456420010 ACRO456420050
N-Formyl-L-proline 97%
N-Formyl-L-proline
Formel: C₆H₉NO₃
Molecular Weight: 143.14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00037321
CAS-Nummer: 13200-83-4

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder or crystalline powder
Infrared spectrum Conforms
Elemental Analysis 9.39 to 10.17 % N
Specific optical rotation -115° to -130° (20°C, 589 nm) (c=0.8 in EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR