Rosuvastatin calcium salt 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
7-{4-(4-Fluorophenyl)-6-isopropyl-2-[methyl(methylsulphonyl)amino]pyrimidin-5-yl}-3,5-dihydroxyhept-6-enoic acid, Calcium (3R,5S,E)-7-(4-(4-fluorophenyl)-6-isopropyl-2-(N-methylmethylsulphonamido)pyrimidin-5-yl)-3,5-dihydroxyhept-6-enoate, Rosuvastatin Calcium, CRESTOR, (3R,5S,6E)-7-[4-(4-fluorophenyl)-6-(1-methylethyl)-2-[N-methyl(n-methylsulphonyl)amino]-5-pyrimidinyl]-3,5-dihydroxy-6-Heptenoic calcium
456230010
456232500
ACRO456230010EA
302
EUR
ACRO456230010
ACRO456232500
Rosuvastatin calcium salt 98%
Rosuvastatin calcium salt
Formel:
C₂₂H₂₇FN₃O₆S·¹/₂Ca Molecular Weight: 501,58 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04112702 CAS-Nummer: 147098-20-2 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...