Diallylmethylamine 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
456340050
456340250
456341000
ACRO456340050EA
47.9
EUR
ACRO456340050
ACRO456340250
ACRO456341000
Diallylmethylamine 98%
Diallylmethylamine
Formel:
C₇H₁₃N Molecular Weight: 111.19 g/mol Siedepunkt: 111 °C (760 mmHg) Dichte: 0.789 g/cm³ Flammpunkt: 7 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00039824 CAS-Nummer: 2424-01-3 UN: 2733 ADR: 3 II |
Spezifikation Testergebnisse
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4290 to 1.4310 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...