3-Amino-1-methyl-1H-pyrazol 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1-Methyl-1H-pyrazol-3-amine, 3-Amino-1-methylpyrazole, 1-Methyl-1H-pyrazol-3-ylamine, 3-Amino-1-methyl-1H-pyrazole

456730050 456730250
ACRO456730050EA 38.4 EUR
ACRO456730050 ACRO456730250
3-Amino-1-methyl-1H-pyrazol 97%
3-Amino-1-methyl-1H-pyrazole
Formel: C₄H₇N₃
Molecular Weight: 97,12 g/mol
Siedepunkt: 93…94 °C (,5 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00466340
CAS-Nummer: 1904-31-0

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow to green to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5400 to 1.5450 (20°C, 589 nm)
Water ≤1.5 %
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR