5-Brom-1H-indazol 97%

Lieferant: Thermo Scientific
Gefahr

456740050 456740250
ACRO456740050EA 78.9 EUR
ACRO456740050 ACRO456740250
5-Brom-1H-indazol 97%
5-Bromo-1H-indazole
Formel: C₇H₅BrN₂
Molecular Weight: 197,03 g/mol
Schmelzpunkt: 123…124 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00839493
CAS-Nummer: 53857-57-1
UN: 2811
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow to brown Crystalline powder or solid
Infrared spectrum Conforms
Melting point 125 to 132 °C
Water ≤1.5 %
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR