Lopinavir 98%
Lieferant: Thermo Scientific
Synonyme:
Koletra, N-((2S,4S,5S)-5-(2-(2,6-Dimethylphenoxy)acetamido)-4-hydroxy-1,6-diphenylhexan-2-yl)-3-methyl-2-(2-oxotetrahydropyrimidin-1(2H)-yl)butanamide, ABT-378, Aluviran, A-157378.0, (S)-N-((2R,4S,5S)-5-(2-(2,6-Dimethylphenoxy)acetamido)-4-hydroxy-1,6-diphenylhexn-2-yl)-3-methyl-2-(2-oxotetrahydropyrimidin-1(2H)-yl)butanamide
456750010
456750050
ACRO456750010EA
138
EUR
ACRO456750010
ACRO456750050
Lopinavir 98%
Lopinavir
Formel:
C₃₇H₄₈N₄O₅ Molecular Weight: 628,82 g/mol Schmelzpunkt: 124…127 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD22628840 CAS-Nummer: 192725-17-0 |
Spezifikation Testergebnisse
Appearance | White to beige Powder |
Infrared spectrum | Conforms |
Water | ≤4 % |
HPLC | ≥97.5 % |
Residue after ignition | ≤0.2 % |
Specific optical rotation | (On anhydrous substance) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...