1-Trifluoromethyl-3,3-dimethyl-1,2-benziodoxole 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3,3-Dimethyl-1-(trifluoromethyl)-1,2-benziodoxole, Togni Reagent

456900010 456900050 456902500
ACRO456900010EA 175 EUR
ACRO456900010 ACRO456900050 ACRO456902500
1-Trifluoromethyl-3,3-dimethyl-1,2-benziodoxole 95%
1-Trifluoromethyl-3,3-dimethyl-1,2-benziodoxole
Formel: C₁₀H₁₀F₃IO
Molecular Weight: 330,09 g/mol
Schmelzpunkt: 75…79 °C
Lagertemperatur: Kühlschrank
MDL: MFCD10567056
CAS-Nummer: 887144-97-0

Bestellen

Spezifikation Testergebnisse

Appearance White to light brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 70 to 79 °C
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR