Fusidic acid 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(Z)-2-((3R,4S,5S,8S,9S,10S,11R,13R,14S,16S)-16-acetoxy-3,11-dihydroxy-4,8,10,14-tetramethyldodecahydro-1H-cyclopenta[a]phenanthren-17(2H,10H,14H)-ylidene)-6-methylhept-5-enoic acid, Fusidine
458460010
458460050
ACRO458460010EA
92.2
EUR
ACRO458460010
ACRO458460050
Fusidic acid 98%
Fusidic acid
Formel:
C₃₁H₄₈O₆ Molecular Weight: 516,72 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00865135 CAS-Nummer: 6990-06-3 |
Spezifikation Testergebnisse
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Water | ≤10 % |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...