3-(Diethylboryl)pyridin 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-(Diethylboryl)pyridine
457750250
457750050
ACRO457750250EA
177
EUR
ACRO457750250
ACRO457750050
3-(Diethylboryl)pyridin 97%
Diethyl(3-pyridyl)borane
Formel:
C₉H₁₄BN Molecular Weight: 147,03 g/mol Schmelzpunkt: 149…151 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012348 CAS-Nummer: 89878-14-8 |
Spezifikation Testergebnisse
Appearance | White to yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 164 to 170 °C |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...