Aripiprazole
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
7-[4-[4-(2,3-Dichlorophenyl)-1-piperazinyl]butoxy]-3,4-dihydro-2(1H)-quinolinone
457990010
457990050
ACRO457990010EA
64.5
EUR
ACRO457990010
ACRO457990050
Aripiprazole
Aripiprazole
Formel:
C₂₃H₂₇Cl₂N₃O₂ Molecular Weight: 448.39 g/mol Schmelzpunkt: 149 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00892072 CAS-Nummer: 129722-12-9 |
Spezifikation Testergebnisse
Appearance | White to beige Powder |
Infrared spectrum | Conforms |
Melting point | 133 to 149 °C |
Loss on drying | ≤0.3 % (105 °C, 3 h) |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...