5-Brom-1,2,3-trimethoxybenzol 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1-Brom-3,4,5-trimethoxybenzol
458130250
458130050
458131000
ACRO458130250EA
117
EUR
ACRO458130250
ACRO458130050
ACRO458131000
5-Brom-1,2,3-trimethoxybenzol 97%
5-Brom-1,2,3-trimethoxybenzol
Formel:
C₉H₁₁BrO₃ Molecular Weight: 247.09 g/mol Schmelzpunkt: 78…82 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017169 CAS-Nummer: 2675-79-8 |
Spezifikation Testergebnisse
Appearance | White to brown Powder or crystalline powder |
Infrared spectrum | Conforms |
Melting point | 75 to 83 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...