4-Amino-3-fluorphenol 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2-Fluoro-4-hydroxyaniline
458310010
458310050
ACRO458310010EA
79
EUR
ACRO458310010
ACRO458310050
4-Amino-3-fluorphenol 98%
4-Amino-3-fluorphenol
Formel:
C₆H₆FNO Molecular Weight: 127,12 g/mol Schmelzpunkt: 137…139 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00077452 CAS-Nummer: 399-95-1 UN: 3077 ADR: 9,III |
Spezifikation Testergebnisse
Appearance | Light yellow to brown Powder |
Infrared spectrum | Conforms |
Melting point | 137 to 141 °C |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...