N-Hexadecyl-N,N-dimethyl-3-ammonio-1-propansulfonat (Sulfobetain-16, SB-16) 98%
Lieferant: Thermo Scientific
Achtung
Synonyme: Palmityldimethyl(3-sulphopropyl)ammonium Hydroxide Inner salt, Hexadecyldimethyl(3-sulphopropyl)ammonium Hydroxide Inner salt, 3-[Dimethyl(palmityl)ammonio]propane-1-sulphonate, Palmityl sulphobetaine, 3-(Hexadecyldimethylammonio)propane-1-sulphonate
458500050 458500250 458501000
ACRO458500050EA 40.7 EUR
ACRO458500050 ACRO458500250 ACRO458501000
N-Hexadecyl-N,N-dimethyl-3-ammonio-1-propansulfonat (Sulfobetain-16, SB-16) 98%
N-Hexadecyl-N,N-dimethyl-3-ammonio-1-propansulfonat (Sulfobetain-16, SB-16)
Formel: C₂₁H₄₅NO₃S
Molecular Weight: 391,6500 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00036911
CAS-Nummer: 2281-11-0
Bestellen
Spezifikation Testergebnisse
Appearance White Powder
Infrared spectrum Conforms
Loss on drying ≤2 %
HPLC ≥97.5 %
Elemental Analysis 63.1 to 65.7 % (C, anhydrous)