Lovastatin 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
6alpha-Methylcompactin, Monacolin K, Mevinolin, (S)-(1S,3R,7S,8S,8aR)-8-(2-((2R,4R)-4-Hydroxy-6-oxotetrahydro-2H-pyran-2-yl)ethyl)-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl 2-methylbutanoate
458830050
458830250
458831000
ACRO458830050EA
74.1
EUR
ACRO458830050
ACRO458830250
ACRO458831000
Lovastatin 98%
Lovastatin
Formel:
C₂₄H₃₆O₅ Molecular Weight: 404,55 g/mol Schmelzpunkt: 175 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00072164 CAS-Nummer: 75330-75-5 |
Spezifikation Testergebnisse
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...