(-)-Cytisin 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
(1R,5S)-1,2,3,4,5,6-Hexahydro-8H-1,5-methanopyrido[1,2-a][1,5]diazocin-8-one, (1R,5S)-3,4,5,6-Tetrahydro-1H-1,5-methanopyrido[1,2-a][1,5]diazocin-8(2H)-one, Cytisine, Cytisinicline
458850010
458852500
ACRO458850010EA
330
EUR
ACRO458850010
ACRO458852500
(-)-Cytisin 98%
(-)-Cytisine
Formel:
C₁₁H₁₄N₂O Molecular Weight: 190,25 g/mol Siedepunkt: 218 °C (3hPa) Schmelzpunkt: 154…156 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00136048 CAS-Nummer: 485-35-8 UN: 2811 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | White to yellow Powder |
Infrared spectrum | Conforms |
Melting point | 152 to 158 °C |
Loss on drying | ≤5 % |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...