Pyrrolidine-3-carboxylic acid 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: H-DL-β-Pro-OH

459110010 459110050
ACRO459110010EA 72 EUR
ACRO459110010 ACRO459110050
Pyrrolidine-3-carboxylic acid 98%
Pyrrolidine-3-carboxylic acid
Formel: C5H9NO2
Molecular Weight: 115.1305 g/mol
Schmelzpunkt: 182…184 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01318699
CAS-Nummer: 59378-87-9

Bestellen

Spezifikation Testergebnisse

Appearance White to light orange Powder or solid
Infrared spectrum Conforms
Elemental Analysis ≥11.86 to ≤12.47 % Nitrogen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR