β-methasone 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
9ɑ-Fluoro-11β,17ɑ,21-trihydroxy-16β-methyl-1,4-pregnadiene-3,20-dione
459070010
459070050
459070250
ACRO459070010EA
39.1
EUR
ACRO459070010
ACRO459070050
ACRO459070250
β-methasone 97%
Betamethasone
Formel:
C₂₂H₂₉FO₅ Molecular Weight: 392.47 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00062969 CAS-Nummer: 378-44-9 |
Spezifikation Testergebnisse
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Melting point | 235 to 245 °C |
Loss on drying | ≤0.5 % |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...