Methyl-L-Lysinat-Dihydrochlorid 98%
Lieferant: Thermo Scientific
Synonyme: H-Lys-OMe·2HCl, H-Lys-OMe.2HCl
459270050 459270250 459271000
ACRO459270050EA 27.4 EUR
ACRO459270050 ACRO459270250 ACRO459271000
Methyl-L-Lysinat-Dihydrochlorid 98%
Methyl-L-Lysinat-Dihydrochlorid
Formel: C₇H₁₆N₂O₂
Molecular Weight: 233,13 g/mol
Schmelzpunkt: 200 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00039067
CAS-Nummer: 26348-70-9
Bestellen
Spezifikation Testergebnisse
Appearance White to off-white Powder
Infrared spectrum Conforms
Specific optical rotation +21° to +25° (20°C, 589 nm) (1% in MeOH)
Titration with HClO4 ≥97.5 %