5-Chloropyrimidine 95%
Lieferant: Thermo Scientific
|
Gefahr
|
459660010
459660050
ACRO459660010EA
88
EUR
ACRO459660010
ACRO459660050
5-Chloropyrimidine 95%
5-Chloropyrimidine
Formel:
C4H3ClN2 Molecular Weight: 114,5300 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03840717 CAS-Nummer: 17180-94-8 UN: 2811 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | White to yellow Crystalline powder or solid |
Infrared spectrum | Conforms |
Water | ≤2.5 % |
GC | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...