1-(3-sulphopropyl)pyridinium hydroxide inner salt 99%
Lieferant: Thermo Scientific
Synonyme:
NDSB 201, 3-(1-Pyridinio)propanesulphonate, 3-(Pyridin-1-ium-1-yl)propane-1-sulphonate
459670500
459672500
ACRO459670500EA
48.6
EUR
ACRO459670500
ACRO459672500
1-(3-sulphopropyl)pyridinium hydroxide inner salt 99%
1-(3-sulphopropyl)pyridinium hydroxide inner salt
Formel:
C₈H₁₁NO₃S Molecular Weight: 201,24 g/mol Schmelzpunkt: 275 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064468 CAS-Nummer: 15471-17-7 |
Spezifikation Testergebnisse
Appearance | White Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤1 % |
Solubility | (10% in water) Clear solution |
HPLC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...