Gatifloxacin 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1-Cyclopropyl-6-fluoro-8-methoxy-7-(3-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid

460230010 460230050 460230250
ACRO460230010EA 42.9 EUR
ACRO460230010 ACRO460230050 ACRO460230250
Gatifloxacin 98%
Gatifloxacin
Formel: C₁₉H₂₂FN₃O₄
Schmelzpunkt: 185…187 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00895399
CAS-Nummer: 112811-59-3

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder
Infrared spectrum Conforms
Loss on drying 4 % to 8 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR