Methyl-β-D-thiogalactoside 98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: TMG, Thiomethyl β-D-galactopyranoside

459450010 459452500
ACRO459450010EA 286 EUR
ACRO459450010 ACRO459452500
Methyl-β-D-thiogalactoside 98%
Methyl-beta-D-thiogalactoside
Formel: C₇H₁₄O₅S
Molecular Weight: 210,25 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00038073
CAS-Nummer: 155-30-6

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 176 to 180 °C
Water ≤1 %
HPLC ≥97.5 %
Specific optical rotation +10° to +12° (20 °C, 589 nm) (5 % in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR