4,6-Pyrimidindiamin 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4,6-Pyrimidinediamine
459800010
459800050
ACRO459800010EA
37.4
EUR
ACRO459800010
ACRO459800050
4,6-Pyrimidindiamin 98%
4,6-Pyrimidindiamin
Formel:
C₄H₆N₄ Molecular Weight: 110,12 g/mol Schmelzpunkt: 271 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00044572 CAS-Nummer: 2434-56-2 |
Spezifikation Testergebnisse
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤1 % |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...