2-Chlor-3-methoxybenzoesäure 97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2-Chlor-m-anissäure
459900010
459900050
ACRO459900010EA
63.1
EUR
ACRO459900010
ACRO459900050
2-Chlor-3-methoxybenzoesäure 97%
2-Chlor-3-methoxybenzoesäure
Formel:
C₈H₇ClO₃ Molecular Weight: 186,59 g/mol Schmelzpunkt: 161…165 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07784399 CAS-Nummer: 33234-36-5 UN: 2811 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | White to off-white Powder or crystalline powder or needles |
Infrared spectrum | Conforms |
Melting point | 160 to 165 °C |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...