Ribavirin 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1H-1,2,4-triazole-3-carboxamide
460480010
460480050
460480250
ACRO460480010EA
87.4
EUR
ACRO460480010
ACRO460480050
ACRO460480250
Ribavirin 98%
Ribavirin
Formel:
C8H12N4O5 Molecular Weight: 244,2047 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00058564 CAS-Nummer: 36791-04-5 |
Spezifikation Testergebnisse
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤0.5 % |
HPLC | ≥97.5 % |
Residue after ign. (sulfated) | ≤0.25 % |
Specific optical rotation | -33° to -37° (20°C, 589 nm) (1 % in water) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...