N4-Benzoylcytosin 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: N-(2-oxo-1,2-dihydropyrimidin-4-yl)benzamide

460540050 460540250
ACRO460540050EA 26.8 EUR
ACRO460540050 ACRO460540250
N4-Benzoylcytosin 98%
N4-Benzoylcytosin
Formel: C₁₁H₉N₃O₂
Molecular Weight: 215,21 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00239434
CAS-Nummer: 26661-13-2

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder or crystalline powder
Infrared spectrum Conforms
Elemental Analysis ≥19.04 to ≤20.01 % Nitrogen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR