Linezolid 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Zyvox, (S)-N-((3-(3-fluoro-4-morpholinophenyl)-2-oxooxazolidin-5-yl)methyl)acetamide
460590010
460592500
ACRO460590010EA
332
EUR
ACRO460590010
ACRO460592500
Linezolid 98%
Linezolid
Formel:
C₁₆H₂₀FN₃O₄ Molecular Weight: 337,4 g/mol Schmelzpunkt: 181…183 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00937825 CAS-Nummer: 165800-03-3 |
Spezifikation Testergebnisse
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤0.5 % |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...