Chlorthalidone 98%

Lieferant: Thermo Scientific

Synonyme: 2-Chloro-5(1-hydroxy-3-oxo-1-isoindolinyl)benzenesulphonamide, Chlortalidone

460932500 460930010
ACRO460932500EA 158 EUR
ACRO460932500 ACRO460930010
Chlorthalidone 98%
Chlorthalidone
Formel: C₁₄H₁₁ClN₂O₄S
Molecular Weight: 338,77 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00036257
CAS-Nummer: 77-36-1

Bestellen

Spezifikation Testergebnisse

Appearance White to beige Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (1 g, 105°C)
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR