Pyrimethamine 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-(4-Chlorophenyl)-6-ethylpyrimidine-2,4-diamine, 5-(4-Chlorophenyl)-6-ethyl-2,4-pyrimidinediamine

461200010 461200050
ACRO461200010EA 31.4 EUR
ACRO461200010 ACRO461200050
Pyrimethamine 98%
Pyrimethamine
Formel: C₁₂H₁₃ClN₄
Molecular Weight: 248,71 g/mol
Schmelzpunkt: 241 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00057350
CAS-Nummer: 58-14-0

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 239 to 243 °C
Loss on drying ≤0.5 % (105 °C, 4 h)
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR