Raltegravir potassium salt
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Raltegravir potassium, Potassium 4-((4-fluorobenzyl)carbamoyl)-1-methyl-2-(2-(5-methyl-1,3,4-oxadiazole-2-carboxamido)propan-2-yl)-6-oxo-1,6-dihydropyrimidin-5-olate
462160010
462162500
ACRO462160010EA
450
EUR
ACRO462160010
ACRO462162500
Raltegravir potassium salt
Raltegravir potassium salt
Formel:
C₂₀H₂₀FKN₆O₅ Molecular Weight: 482,51 g/mol Schmelzpunkt: 155…157 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD12031642 CAS-Nummer: 871038-72-1 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | ≤1 % |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...