Anastrozole 98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: alpha,alpha,alpha',alpha'-Tetramethyl-5-(1H-1,2,4-triazol-1-ylmethyl)-1,3-benzenediacetonitrile

460970010 460972500
ACRO460970010EA 479 EUR
ACRO460970010 ACRO460972500
Anastrozole 98%
Anastrozole
Formel: C₁₇H₁₉N₅
Molecular Weight: 293,37 g/mol
Schmelzpunkt: 81…82 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00866298
CAS-Nummer: 120511-73-1

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 82 to 86 °C
Water ≤0.3 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR