Azaperone

Lieferant: Thermo Scientific
Gefahr

Synonyme: 4'-Fluoro-4-[4-(2-pyridyl)-1-piperazinyl]butyrophenone, 1-(4-Fluorophenyl)-4-(4-(2-pyridinyl)-1-piperazinyl)-1-butanone

460830010 460830050 460830250
ACRO460830010EA 89.3 EUR
ACRO460830010 ACRO460830050 ACRO460830250
Azaperone
Azaperone
Formel: C₁₉H₂₂FN₃₀
Molecular Weight: 689,58 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00866692
CAS-Nummer: 1649-18-9
UN: 2811
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 92 to 95 °C
Loss on drying ≤0.5 % (60 °C, 4 h) (vacuum)
Titration with HClO4 99 to 101 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR